Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002325.1 | 1 | 35 | |||
GenBank mRNA | AF375472.1 | - | 1 | 35 | ||
AJ609429.1 | - | 1 | 35 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN299976.1 | - | 1 | 222 | Chr. 3 - 39408204 - 39410850 [+] on Build GRCh38 |