Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PET100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033242.1 | 4 | 255 | |||
GenBank mRNA | AK124717.1 | - | 1 | 1491 | ||
CB119056.1 | - | 4 | 246 |