Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKFZP434L187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026771.1 | 1, 2 | 81 | |||
GenBank mRNA | BC033224.1 | - | 1, 2 | 424 | ||
BC130408.1 | - | 1, 2 | 81 | |||
BC130410.1 | - | 1, 2 | 81 |
Gene Symbol: LOC105370751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125576.1 | - | 1, 2 | 203 |