Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015356.2 | 5 | 601 | |||
GenBank mRNA | AY312364.1 | - | 6 | 749 | ||
AY312365.1 | - | LIT3317 AAA1 |
5 | 599 | ||
BC031961.1 | - | 6 | 727 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436945.4 | - | 5 | 598 | Chr. 7 - 34718862 - 34834328 [-] on Build GRCh38 |