Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015356.2 | 5 | 1053 | |||
GenBank mRNA | AY312364.1 | - | 6 | 1201 | ||
AY312365.1 | - | LIT3317 AAA1 |
5 | 1051 | ||
BC031961.1 | - | 6 | 1179 |