Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002976.1 | 1 | 96 | |||
GenBank mRNA | AJ609466.1 | - | 1 | 96 |
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034611.1 | - | 2 | 887 | ||
BC064589.1 | - | 3 | 602 | |||
BC107730.1 | - | 1 | 432 |