Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN8P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002830.1 | 1 | 285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU159632.1 | - | 1 | 282 | Chr. 1 - 89994020 - 89994963 [-] on Build GRCh38 |