Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf62-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024622.1 | 4 | 720 | |||
NR_024623.1 | 4 | 708 | ||||
GenBank mRNA | AK096601.1 | - | 4 | 709 | ||
BC034794.1 | - | 2 | 434 | |||
BC117397.1 | - | 4 | 602 | |||
BC117399.1 | - | 4 | 614 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV717800.1 | - | 4 | 459 | Chr. 21 - 32772174 - 32797446 [+] on Build GRCh38 |