Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTFR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024369.1 | 2 | 809 | |||
GenBank mRNA | BC033987.1 | - | 3 | 921 | ||
BC038740.1 | - | 5 | 1242 | |||
BC042806.1 | - | 2 | 810 |