Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC202181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026921.1 | 2 | 1012 | |||
GenBank mRNA | Y13871.1 | - | 1 | 719 |
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308195.1 | NP_001295124.1 | 2 | 1042 | ||
NM_001308196.1 | NP_001295125.1 | 4 | 1250 | |||
XM_011534553.2 | XP_011532855.1 | 3 | 5047 | |||
XM_011534554.2 | XP_011532856.1 | 3 | 919 | |||
XM_011534556.2 | XP_011532858.1 | 2 | 4916 | |||
XM_017009454.1 | XP_016864943.1 | 2 | 850 | |||
XM_017009455.1 | XP_016864944.1 | 3 | 1160 | |||
XM_017009456.1 | XP_016864945.1 | 3 | 1056 | |||
GenBank mRNA | AK126204.1 | - | 1 | 1578 | ||
AK299336.1 | - | 2 | 1002 | |||
BC037298.1 | - | 4 | 1251 | 4823991 | ||
BX390643.2 | - | 1 | 125 | |||
KF151170.1 | - | 2 | 910 |