Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003003.2 | 1 | 209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB997907.1 | - | 1 | 234 | Chr. 18 - 49814023 - 49814677 [+] on Build GRCh38 |