Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033361.1 | 1 | 77 | |||
XM_006713552.3 | XP_006713615.1 | 1 | 122 | |||
GenBank mRNA | AK124671.1 | - | 1 | 78 |