Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252507.2 | NP_001239436.1 | 2 | 773 | ||
NM_001282463.1 | NP_001269392.1 | 3 | 872 | |||
NM_001289139.1 | NP_001276068.1 | 3 | 866 | |||
XM_017021469.1 | XP_016876958.1 | 2 | 2105 | |||
GenBank mRNA | AB621830.1 | - | 2 | 735 | ||
AK001590.1 | - | 2 | 717 | |||
BC043593.1 | - | 2 | 756 |