Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184906.1 | NP_001171835.1 | 14 | 9448 | ||
NM_032875.2 | NP_116264.2 | 15 | 9544 | |||
XM_005257746.3 | XP_005257803.2 | 15 | 9735 | |||
XM_005257747.3 | XP_005257804.2 | 14 | 9516 | |||
GenBank mRNA | AL117623.1 | - | 1 | 606 | ||
BC042012.1 | - | 1 | 816 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ719214.1 | - | 1 | 555 | Chr. 17 - 39253150 - 39254035 [-] on Build GRCh38 |