Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400487.1 | - | LIT2225 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2 | 153 | |
AL832758.1 | - | 14 | 1843 | |||
BC080646.1 | - | 14 | 1881 | |||
BX648788.1 | - | 4 | 403 |