Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF041080.1 | - | LIT2190 HERC2P2 |
8 | 1004 |