Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273481.2 | XP_005273538.1 | 8 | 5688 | ||
GenBank mRNA | AK090522.1 | - | 1 | 1776 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK091357.1 | - | 1 | 952 | Chr. 8 - 17224967 - 17227277 [-] on Build GRCh38 |