Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022805.1 | - | 1 | 2464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | N99347.1 | - | 1 | 68 | Chr. 9 - 21324052 - 21326339 [-] on Build GRCh38 |