Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530934.2 | XP_011529236.1 | 8 | 15878 | ||
XM_017029473.1 | XP_016884962.1 | 8 | 14848 | |||
XM_017029474.1 | XP_016884963.1 | 8 | 10990 | |||
XM_017029475.1 | XP_016884964.1 | 4 | 14233 | |||
XM_017029476.1 | XP_016884965.1 | 4 | 10350 | |||
XM_017029477.1 | XP_016884966.1 | 4 | 15264 | |||
GenBank mRNA | AK056217.1 | - | 1 | 99 |