Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311649.1 | - | 2 | 719 | ||
BC011026.1 | - | 2 | 752 | |||
BC014548.1 | - | 3 | 821 | |||
BC039236.1 | - | 3 | 744 | |||
BC047894.1 | - | 3 | 850 | |||
BC063120.1 | - | 3 | 845 |