Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136638.1 | 2 | 1002 | |||
NR_136639.1 | 2 | 706 | ||||
NR_136640.1 | 2 | 271 | ||||
NR_136641.1 | 2 | 450 | ||||
GenBank mRNA | AA401699.1 | - | 2 | 257 | ||
BC008048.2 | - | 2 | 1001 | |||
BC033986.1 | - | 2 | 672 |