Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080512.2 | NP_001073981.1 | 21 | 3926 | ||
XM_005270169.4 | XP_005270226.1 | 20 | 6924 | |||
XM_011540185.2 | XP_011538487.1 | 22 | 4172 | |||
XM_011540188.2 | XP_011538490.2 | 21 | 4057 | |||
XM_011540189.2 | XP_011538491.1 | 20 | 3823 | |||
XM_011540190.2 | XP_011538492.1 | 20 | 3780 | |||
XM_011540191.2 | XP_011538493.1 | 18 | 3642 | |||
XM_017016677.1 | XP_016872166.1 | 22 | 10560 | |||
XM_017016678.1 | XP_016872167.1 | 23 | 4077 | |||
GenBank mRNA | AK130049.1 | - | 1 | 702 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA401855.1 | - | 1 | 194 | Chr. 10 - 58829699 - 58830341 [+] on Build GRCh38 |