Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU6F2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023033.1 | - | 6 | 712 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA705054.1 | - | 5 | 495 | Chr. 7 - 38981468 - 39013551 [-] on Build GRCh38 |