Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718225.2 | XP_006718288.1 | 7 | 2056 | ||
GenBank mRNA | CR627308.1 | - | 1 | 6203 |