Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250045.2 | XP_005250102.1 | 19 | 3363 | ||
XM_017012558.1 | XP_016868047.1 | 20 | 3483 | |||
XM_017012559.1 | XP_016868048.1 | 19 | 3329 | |||
GenBank mRNA | AK124257.1 | - | 1 | 934 |