Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316341.1 | NP_001303270.1 | 13 | 4494 | ||
NM_002816.4 | NP_002807.1 | 11 | 4210 | |||
NM_174871.3 | NP_777360.1 | 10 | 4150 | |||
GenBank mRNA | AB003103.1 | - | 12 | 3353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095141.1 | - | 1823 | Chr. - - [-] on Build |