Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 61 | 6798 | ||
XM_006717308.2 | XP_006717371.1 | 60 | 6812 | |||
XM_006717310.3 | XP_006717373.1 | 59 | 4611 | |||
XM_011519138.2 | XP_011517440.1 | 61 | 6430 | |||
XM_011519140.1 | XP_011517442.1 | 60 | 6812 | |||
XM_011519145.2 | XP_011517447.1 | 48 | 4040 | |||
GenBank mRNA | AB058773.1 | - | 58 | 5288 | ||
AK021957.1 | - | 3 | 1543 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR605514.1 | - | 1 | 508 | Chr. 9 - 114310997 - 114312467 [+] on Build GRCh38 |