Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282670.1 | NP_001269599.1 | 1 | 36 | ||
GenBank mRNA | AK126870.1 | - | 1 | 36 |
Gene Symbol: LOC105378592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003075.1 | XP_016858564.1 | 1 | 2509 | ||
XM_017003076.1 | XP_016858565.1 | 1 | 2509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024162.1 | - | 1 | 1499 | Chr. 1 - 2211921 - 2214181 [-] on Build GRCh38 |