Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110512.1 | 2 | 200 | |||
GenBank mRNA | AK055475.1 | - | 2 | 461 | ||
DA365834.1 | - | 2 | 175 |