Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 14 | 4815 | ||
NM_144966.5 | NP_659403.4 | 38 | 9934 | |||
XM_005251382.3 | XP_005251439.1 | 37 | 10182 | |||
XM_005251384.4 | XP_005251441.1 | 16 | 5228 | |||
XM_006716729.3 | XP_006716792.1 | 17 | 5473 | |||
XM_011517758.2 | XP_011516060.1 | 14 | 4864 | |||
XM_017014316.1 | XP_016869805.1 | 37 | 10209 | |||
XM_017014317.1 | XP_016869806.1 | 38 | 9752 | |||
XM_017014318.1 | XP_016869807.1 | 38 | 10507 | |||
XM_017014319.1 | XP_016869808.1 | 39 | 10411 | |||
XM_017014320.1 | XP_016869809.1 | 38 | 10338 | |||
XM_017014321.1 | XP_016869810.1 | 37 | 10001 | |||
XM_017014322.1 | XP_016869811.1 | 37 | 9795 | |||
XM_017014323.1 | XP_016869812.1 | 37 | 9468 | |||
XM_017014324.1 | XP_016869813.1 | 36 | 10059 | |||
XM_017014325.1 | XP_016869814.1 | 36 | 10093 | |||
XM_017014326.1 | XP_016869815.1 | 36 | 9025 | |||
XM_017014327.1 | XP_016869816.1 | 33 | 8394 | |||
GenBank mRNA | AK054690.1 | - | 1 | 1719 |