Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 14 | 3729 | ||
NM_144966.5 | NP_659403.4 | 38 | 8848 | |||
XM_005251382.3 | XP_005251439.1 | 37 | 9096 | |||
XM_005251384.4 | XP_005251441.1 | 16 | 4142 | |||
XM_006716729.3 | XP_006716792.1 | 17 | 4387 | |||
XM_011517758.2 | XP_011516060.1 | 14 | 3778 | |||
XM_017014316.1 | XP_016869805.1 | 37 | 9123 | |||
XM_017014317.1 | XP_016869806.1 | 38 | 8666 | |||
XM_017014318.1 | XP_016869807.1 | 38 | 9421 | |||
XM_017014319.1 | XP_016869808.1 | 39 | 9325 | |||
XM_017014320.1 | XP_016869809.1 | 38 | 9252 | |||
XM_017014321.1 | XP_016869810.1 | 37 | 8915 | |||
XM_017014322.1 | XP_016869811.1 | 37 | 8709 | |||
XM_017014323.1 | XP_016869812.1 | 37 | 8382 | |||
XM_017014324.1 | XP_016869813.1 | 36 | 8973 | |||
XM_017014325.1 | XP_016869814.1 | 36 | 9007 | |||
XM_017014326.1 | XP_016869815.1 | 36 | 7939 | |||
XM_017014327.1 | XP_016869816.1 | 33 | 7308 | |||
GenBank mRNA | AK054690.1 | - | 1 | 633 |