Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301085.1 | NP_001288014.1 | 8 | 7302 | ||
NM_006775.2 | NP_006766.1 | 8 | 7326 | |||
NM_206853.2 | NP_996735.1 | 7 | 11999 | |||
NM_206854.2 | NP_996736.1 | 7 | 13278 | |||
NM_206855.2 | NP_996737.1 | 6 | 14225 | |||
XM_011536258.2 | XP_011534560.1 | 8 | 6830 | |||
XM_011536260.2 | XP_011534562.1 | 7 | 7224 | |||
GenBank mRNA | AK096701.1 | - | 1 | 2216 |