Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278119.1 | NP_001265048.1 | 7 | 6082 | ||
NM_001278121.1 | NP_001265050.1 | 7 | 6029 | |||
NM_001278122.1 | NP_001265051.1 | 6 | 6002 | |||
NM_006298.3 | NP_006289.2 | 6 | 5988 | |||
XM_011514870.2 | XP_011513172.1 | 6 | 6077 | |||
XM_011514873.2 | XP_011513175.1 | 7 | 6188 | |||
XM_011514874.2 | XP_011513176.1 | 5 | 5485 | |||
XM_017011266.1 | XP_016866755.1 | 6 | 5992 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR606559.1 | - | 1 | 267 | Chr. 6 - 28157790 - 28159467 [+] on Build GRCh38 |