Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160451.1 | - | 3 | 627 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707685.1 | - | 3 | 623 | Chr. 9 - 75232055 - 75233315 [+] on Build GRCm38 |