Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084703.1 | - | 2 | 2367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB478650.2 | - | 1 | 614 | Chr. 10 - 13381405 - 13382080 [-] on Build GRCm38 |