Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A030001D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003910.1 | - | 1 | 37 | ||
AK037147.1 | - | 1 | 53 | |||
AK049852.1 | - | 1 | 39 |
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CO039349.1 | - | 1 | 403 |