Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125727.1 | 6 | 1690 | |||
NR_125728.1 | 5 | 1537 | ||||
NR_125729.1 | 5 | 1492 | ||||
NR_132994.1 | 6 | 2191 | ||||
NR_132995.1 | 6 | 1645 | ||||
NR_132996.1 | 4 | 1464 | ||||
GenBank mRNA | AK023611.1 | - | 2 | 2585 | ||
AL162041.1 | - | 1 | 637 | |||
BC011838.2 | - | 6 | 1579 | |||
BC017330.2 | - | 5 | 1420 | |||
BC030061.1 | - | 5 | 1420 | |||
BC065547.1 | - | 6 | 2148 | |||
CA334877.1 | - | 89 |