Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126159.1 | 2 | 437 | |||
GenBank mRNA | AK127362.1 | - | 1 | 587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416725.1 | - | 2 | 464 | Chr. 11 - 131662131 - 131663583 [-] on Build GRCh38 |