siRNA Details
Gene Symbol: LOC101928323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125400.1 | 1 | 249 | |||
GenBank mRNA | HY122338.1 | - | 1 | 250 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434363.1 | - | 1 | 187 | Chr. 3 - 42632568 - 42654326 [-] on Build GRCh38 |
