Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE40-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037903.3 | 3 | 1023 | |||
NR_125914.1 | 3 | 982 | ||||
NR_125915.1 | 4 | 927 | ||||
NR_125916.1 | 3 | 787 | ||||
GenBank mRNA | AK311159.1 | - | 3 | 982 | ||
AK311646.1 | - | 3 | 999 |