Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDNF-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002832.2 | 3, 4 | 223 | |||
NR_033314.1 | 3, 4 | 223 | ||||
GenBank mRNA | AY054391.4 | - | 3, 4 | 253 | ||
AY054403.4 | - | 3, 4 | 253 | |||
AY054405.4 | - | 3, 4 | 253 | |||
EF689024.1 | - | 1, 2 | 62 | |||
EF689025.1 | - | 1, 2 | 62 | |||
EF689026.1 | - | 1, 2 | 62 | |||
EF689027.1 | - | 1, 2 | 62 | |||
EF689028.1 | - | 1, 2 | 62 | |||
EF689029.1 | - | 1, 2 | 62 | |||
EF689030.1 | - | 3, 4 | 228 | |||
EF689031.1 | - | 3, 4 | 228 | |||
EF689032.1 | - | 3, 4 | 228 | |||
EF689033.1 | - | 3, 4 | 228 | |||
EF689034.1 | - | 3, 4 | 228 | |||
EF689035.1 | - | 3, 4 | 228 | |||
EF689042.1 | - | 3, 4 | 228 |