Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002806.2 | 3 | 681 | |||
GenBank mRNA | AK093650.1 | - | 1 | 477 | ||
AK126217.1 | - | 3 | 681 |