Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125823.1 | 2, 3 | 457 | |||
GenBank mRNA | BX094193.1 | - | 2, 3 | 229 |