Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105370489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135258.1 | 2 | 197 | |||
GenBank mRNA | HY010266.1 | - | 2 | 198 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555966.1 | - | 2 | 114 | Chr. 14 - 50821880 - 50823501 [+] on Build GRCh38 |