Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927972 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125846.1 | 1 | 158 | |||
NR_125847.1 | 1 | 158 | ||||
NR_125848.1 | 1 | 158 | ||||
GenBank mRNA | CB046455.1 | - | 1 | 125 | ||
DB135850.1 | - | 1 | 158 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000602674.1 | - | 1 | 116 | Chr. 6 - 5451683 - 5458033 [-] on Build GRCh38 |