Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126018.1 | 1 | 23 | |||
NR_126019.1 | 1 | 23 | ||||
GenBank mRNA | BC042032.1 | - | 1 | 23 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000606761.1 | - | 1 | 23 | Chr. 6 - 5029972 - 5043449 [+] on Build GRCh38 |