Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHLAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104319.1 | 5 | 874 | |||
NR_104320.1 | 4 | 805 | ||||
NR_104321.1 | 3 | 538 | ||||
NR_104322.1 | 7 | 1113 | ||||
NR_104323.1 | 6 | 1012 | ||||
NR_104324.1 | 5 | 906 | ||||
NR_104325.1 | 6 | 975 | ||||
GenBank mRNA | JX117418.1 | - | 5 | 874 | ||
JX117419.1 | - | 4 | 805 | |||
JX117420.1 | - | 3 | 538 | |||
JX117421.1 | - | 7 | 1113 | |||
JX117422.1 | - | 6 | 1012 | |||
JX117423.1 | - | 5 | 906 | |||
JX117424.1 | - | 6 | 975 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000629145.1 | - | 874 | Chr. - - [-] on Build |