Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039992.2 | 5 | 385 | |||
GenBank mRNA | BC042069.1 | - | 2 | 153 | ||
BX538073.1 | - | 5 | 362 | |||
BX538140.1 | - | 4 | 334 |