Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054073.2 | NP_473414.1 | 8 | 950 | ||
GenBank mRNA | AF294348.1 | - | 8 | 918 | ||
AK006265.1 | - | 8 | 951 | |||
BC125597.1 | - | 7 | 868 | 40130112 | ||
BC125599.1 | - | 7 | 868 | 40130114 |