Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001981.3 | NP_001001981.2 | 3 | 2279 | ||
NM_001136226.1 | NP_001129698.1 | 4 | 2286 | |||
GenBank mRNA | AK029972.1 | - | 4 | 2289 | ||
AK046254.1 | - | 1 | 195 | |||
AY316161.1 | - | 3 | 2066 | |||
AY316162.1 | - | 1 | 314 | |||
BC039778.1 | - | 1 | 1026 | |||
BC141102.1 | - | 2 | 1971 | 9055714 |